Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275297.1 | NP_001262226.1 | 6 - 7 | 2918 | 69 | |
NM_001300221.1 | NP_001287150.1 | 5 - 6 | 3059 | 69 | ||
NM_141169.3 | NP_649426.3 | 4 - 5 | 2874 | 69 | ||
NM_168975.2 | NP_730762.2 | 5 - 6 | 2843 | 69 | ||
NM_168976.2 | NP_730763.2 | 5 - 6 | 2774 | 69 | ||
GenBank mRNA | BT128821.1 | - | 5 - 6 | 2813 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |