Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032236.3 | NP_001027407.2 | 3 - 4 | 655 | 57 | |
NM_001032237.3 | NP_001027408.1 | 3 - 4 | 974 | 57 | ||
NM_001103817.2 | NP_001097287.1 | 3 - 4 | 974 | 57 | ||
NM_001259351.2 | NP_001246280.1 | 3 - 4 | 974 | 57 | ||
NM_078988.4 | NP_523712.3 | 3 - 4 | 493 | 57 | ||
NM_165904.3 | NP_725171.1 | 3 - 4 | 974 | 57 | ||
NM_165905.4 | NP_725172.2 | 3 - 4 | 493 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |