Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165904.3 | NP_725171.1 | 5 - 6 | 2388 | 82 | |
NM_165905.4 | NP_725172.2 | 5 - 6 | 1907 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |