Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp74F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079408.6 | NP_524132.1 | 5 - 6 | 658 | 66 | |
NM_168754.3 | NP_730317.1 | 4 - 5 | 553 | 66 | ||
GenBank mRNA | AY071260.1 | - | 4 - 5 | 545 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |