Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: verm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169998.3 | NP_001163469.1 | 4 - 5 | 351 | 95 | |
NM_168808.4 | NP_730441.1 | 3 - 4 | 279 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |