Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275133.1 | NP_001262062.1 | 2 - 3 | 458 | 88 | |
NM_168811.2 | NP_730444.1 | 2 - 3 | 773 | 88 | ||
GenBank mRNA | BT058041.1 | - | 2 - 3 | 458 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |