Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir76a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104177.3 | NP_001097647.3 | 7 - 8 | 1458 | 80 | |
NM_001144510.3 | NP_001137982.1 | 5 - 6 | 1202 | 80 | ||
NM_140891.4 | NP_649148.2 | 5 - 6 | 1189 | 80 | ||
NM_168817.3 | NP_730452.2 | 6 - 7 | 1228 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |