Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140999.3 | NP_649256.1 | 3 - 4 | 1561 | 107 | |
NM_168872.1 | NP_730566.1 | 3 - 4 | 1138 | 107 | ||
GenBank mRNA | AF099185.1 | - | 3 - 4 | 1527 | 107 | |
AF247709.1 | - | 3 - 4 | 1138 | 107 | ||
BT028820.2 | - | 3 - 4 | 1087 | 107 | ||
BT031315.1 | - | 3 - 4 | 1552 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |