Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273843.1 | NP_001260772.1 | 6 - 7 | 838 | 84 | |
NM_001273844.1 | NP_001260773.1 | 6 - 7 | 965 | 84 | ||
NM_001273846.1 | NP_001260775.1 | 6 - 7 | 838 | 84 | ||
NM_136441.4 | NP_610285.5 | 8 - 9 | 1304 | 84 | ||
NM_165526.3 | NP_724560.1 | 5 - 6 | 1037 | 84 | ||
NM_165527.3 | NP_724561.2 | 6 - 7 | 876 | 84 | ||
NM_165528.3 | NP_724562.2 | 8 - 9 | 1825 | 84 | ||
NM_165529.3 | NP_724563.2 | 6 - 7 | 784 | 84 | ||
NM_165530.3 | NP_724564.2 | 6 - 7 | 965 | 84 | ||
NM_165531.3 | NP_724565.3 | 8 - 9 | 1180 | 84 | ||
NM_165532.3 | NP_724566.3 | 8 - 9 | 1084 | 84 | ||
NM_206050.3 | NP_995772.1 | 6 - 7 | 838 | 84 | ||
GenBank mRNA | BT003248.1 | - | 5 - 6 | 1046 | 84 | |
BT003252.1 | - | 6 - 7 | 933 | 84 | ||
BT044325.1 | - | 6 - 7 | 837 | 84 | ||
BT126071.1 | - | 8 - 9 | 1331 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |