Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136616.5 | NP_610460.1 | 2 - 3 | 1211 | 63 | |
NM_206061.2 | NP_995783.1 | 2 - 3 | 648 | 63 | ||
GenBank mRNA | AY060801.1 | - | 2 - 3 | 618 | 63 | |
BT050564.1 | - | 2 - 3 | 1179 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |