Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273912.1 | NP_001260841.1 | 8 - 9 | 3338 | 74 | |
NM_001273913.2 | NP_001260842.1 | 8 - 9 | 2102 | 74 | ||
NM_001382026.1 | NP_001369068.1 | 6 - 7 | 1925 | 74 | ||
NM_001382027.1 | NP_001369069.1 | 7 - 8 | 2024 | 74 | ||
NM_001382028.1 | NP_001369070.1 | 6 - 7 | 1943 | 74 | ||
NM_001382029.1 | NP_001369071.1 | 7 - 8 | 2042 | 74 | ||
NM_057670.5 | NP_477018.1 | 7 - 8 | 2042 | 74 | ||
NM_057671.5 | NP_477019.1 | 7 - 8 | 2024 | 74 | ||
NM_057672.5 | NP_477020.1 | 8 - 9 | 2425 | 74 | ||
NM_057673.5 | NP_477021.1 | 7 - 8 | 2024 | 74 | ||
NM_206067.4 | NP_995789.1 | 7 - 8 | 2153 | 74 | ||
GenBank mRNA | BT133393.1 | - | 7 - 8 | 2019 | 74 | |
X83527.1 | - | 2006 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |