Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058159.5 | NP_477507.1 | 8 - 9 | 1132 | 74 | |
NM_134303.4 | NP_599130.1 | 8 - 9 | 1033 | 74 | ||
NM_166178.3 | NP_725586.1 | 8 - 9 | 1069 | 74 | ||
NM_206134.2 | NP_995856.2 | 8 - 9 | 992 | 74 | ||
GenBank mRNA | AY095026.1 | - | 8 - 9 | 1062 | 74 | |
AY128437.1 | - | 8 - 9 | 977 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |