Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169707.2 | NP_001163178.1 | 3 - 4 | 797 | 73 | |
NM_137400.4 | NP_611244.1 | 3 - 4 | 708 | 73 | ||
NM_206157.2 | NP_995879.1 | 3 - 4 | 701 | 73 | ||
GenBank mRNA | AF181625.1 | - | 3 - 4 | 691 | 73 | |
BT083389.1 | - | 3 - 4 | 784 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |