Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144353.4 | NP_652610.1 | 6 - 7 | 1272 | 65 | |
GenBank mRNA | AF222864.1 | - | 6 - 7 | 1248 | 65 | |
BT023840.1 | - | 6 - 7 | 1248 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |