Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc62B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206236.3 | NP_995958.2 | 1 - 2 | 2175 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |