Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CycJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079179.3 | NP_523903.1 | 1 - 2 | 197 | 69 | |
NM_206267.2 | NP_995989.1 | 1 - 2 | 197 | 69 | ||
GenBank mRNA | AY058290.1 | - | 1 - 2 | 211 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |