Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TrxT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131984.1 | NP_572212.1 | 2 - 3 | 564 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |