Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297969.1 | NP_001284898.1 | 1 - 2 | 838 | 97 | |
NM_132004.3 | NP_572232.2 | 1 - 2 | 838 | 97 | ||
GenBank mRNA | BT025193.1 | - | 1 - 2 | 807 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |