Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vanin-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132069.3 | NP_572297.1 | 1 - 2 | 91 | 93 | |
GenBank mRNA | AJ276261.1 | - | 1 - 2 | 80 | 93 | |
AY052034.1 | - | 1 - 2 | 91 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |