Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136073.3 | NP_609917.2 | 1 - 2 | 685 | 66 | |
GenBank mRNA | BT021951.1 | - | 1 - 2 | 669 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |