Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274391.1 | NP_001261320.1 | 2 - 3 | 647 | 71 | |
NM_138984.4 | NP_620469.2 | 2 - 3 | 647 | 71 | ||
GenBank mRNA | BT053678.1 | - | 2 - 3 | 645 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |