Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: He | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298991.1 | NP_001285920.1 | 1 - 2 | 158 | 130 | |
NM_144477.3 | NP_652734.2 | 1 - 2 | 158 | 130 | ||
GenBank mRNA | AF426744.1 | - | 1 - 2 | 158 | 130 | |
BT099873.1 | - | 1 - 2 | 124 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |