Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080084.3 | NP_524823.1 | 4 - 5 | 1754 | 119 | |
NM_166145.2 | NP_725531.1 | 4 - 5 | 1754 | 119 | ||
GenBank mRNA | AF212367.1 | - | 4 - 5 | 1717 | 119 | |
AF212369.1 | - | 4 - 5 | 1717 | 119 | ||
BT150099.1 | - | 4 - 5 | 1710 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |