Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079482.4 | NP_524206.1 | 2 - 3 | 479 | 81 | |
NM_168938.2 | NP_730689.1 | 2 - 3 | 182 | 81 | ||
GenBank mRNA | BT028846.2 | - | 1 - 2 | 96 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |