Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259879.2 | NP_001246808.1 | 4 - 5 | 872 | 70 | |
NM_001275047.1 | NP_001261976.2 | 2 - 3 | 657 | 70 | ||
NM_140673.5 | NP_648930.1 | 4 - 5 | 883 | 70 | ||
NM_170635.3 | NP_730212.1 | 3 - 4 | 827 | 70 | ||
NM_206387.3 | NP_996109.1 | 4 - 5 | 771 | 70 | ||
NM_206388.3 | NP_996110.1 | 4 - 5 | 995 | 70 | ||
GenBank mRNA | AJ418713.1 | - | 4 - 5 | 865 | 70 | |
AY069696.1 | - | 4 - 5 | 865 | 70 | ||
BT009969.1 | - | 4 - 5 | 880 | 70 | ||
BT100317.1 | - | 3 - 4 | 825 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |