Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259940.1 | NP_001246869.1 | 1 - 2 | 146 | 92 | |
NM_206416.2 | NP_996138.2 | 1 - 2 | 146 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |