Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PsGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272258.1 | NP_001259187.1 | 15 - 16 | 8675 | 68 | |
NM_001272259.1 | NP_001259188.1 | 14 - 15 | 8310 | 68 | ||
NM_130645.6 | NP_570001.5 | 15 - 16 | 8392 | 68 | ||
NM_206610.5 | NP_996333.4 | 17 - 18 | 8509 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |