Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnp4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103413.3 | NP_001096883.3 | 6 - 7 | 2353 | 71 | |
NM_001103414.3 | NP_001096884.1 | 7 - 8 | 2383 | 71 | ||
NM_078492.4 | NP_511047.2 | 6 - 7 | 2560 | 71 | ||
NM_206630.3 | NP_996353.1 | 7 - 8 | 2479 | 71 | ||
GenBank mRNA | AY061395.1 | - | 6 - 7 | 2574 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |