Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298385.1 | NP_001285314.1 | 2 - 3 | 1203 | 65 | |
NM_001298386.1 | NP_001285315.1 | 3 - 4 | 985 | 65 | ||
NM_078637.3 | NP_523361.1 | 2 - 3 | 566 | 65 | ||
NM_167480.2 | NP_727925.1 | 3 - 4 | 683 | 65 | ||
NM_167481.3 | NP_727926.2 | 2 - 3 | 868 | 65 | ||
NM_206759.2 | NP_996482.1 | 4 - 5 | 789 | 65 | ||
GenBank mRNA | AJ012609.1 | - | 2 - 3 | 505 | 65 | |
BT044449.1 | - | 2 - 3 | 566 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |