Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272736.2 | NP_001259665.2 | 12 - 13 | 2399 | 85 | |
NM_001272737.1 | NP_001259666.2 | 12 - 13 | 3174 | 85 | ||
NM_078669.5 | NP_523393.3 | 12 - 13 | 2464 | 85 | ||
NM_167592.4 | NP_728120.1 | 12 - 13 | 1998 | 85 | ||
GenBank mRNA | BT050432.1 | - | 12 - 13 | 1998 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |