Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272746.1 | NP_001259675.1 | 2 - 3 | 719 | 65 | |
NM_080354.4 | NP_525093.1 | 2 - 3 | 524 | 65 | ||
NM_206778.3 | NP_996501.1 | 2 - 3 | 586 | 65 | ||
NM_206779.3 | NP_996502.1 | 3 - 4 | 449 | 65 | ||
GenBank mRNA | BT133347.1 | - | 2 - 3 | 522 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |