Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258868.3 | NP_001245797.1 | 4 - 5 | 1351 | 83 | |
NM_001258869.3 | NP_001245798.1 | 4 - 5 | 1234 | 83 | ||
NM_001298597.1 | NP_001285526.1 | 4 - 5 | 1206 | 83 | ||
NM_134617.5 | NP_608461.1 | 3 - 4 | 1420 | 83 | ||
GenBank mRNA | BT099938.1 | - | 3 - 4 | 1406 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |