Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ent1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272891.1 | NP_001259820.1 | 3 - 4 | 691 | 88 | |
NM_134675.5 | NP_608519.2 | 3 - 4 | 709 | 88 | ||
NM_164381.2 | NP_722628.1 | 2 - 3 | 758 | 88 | ||
GenBank mRNA | AY061261.1 | - | 2 - 3 | 752 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |