Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134713.4 | NP_608557.4 | 4 - 5 | 617 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |