Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chinmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201751.2 | NP_001188680.1 | 5 - 6 | 3204 | 73 | |
NM_001258910.2 | NP_001245839.1 | 5 - 6 | 3204 | 73 | ||
NM_134766.6 | NP_608610.1 | 4 - 5 | 1823 | 73 | ||
NM_164428.3 | NP_722717.2 | 6 - 7 | 1921 | 73 | ||
NM_164429.3 | NP_722718.1 | 6 - 7 | 2973 | 73 | ||
NM_164430.4 | NP_722719.1 | 5 - 6 | 2903 | 73 | ||
GenBank mRNA | AY060464.1 | - | 6 - 7 | 1930 | 73 | |
BT050405.1 | - | 5 - 6 | 2902 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |