Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273039.1 | NP_001259968.1 | 8 - 9 | 4654 | 68 | |
NM_134871.3 | NP_608715.2 | 8 - 9 | 4705 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |