Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134983.3 | NP_608827.1 | 1 - 2 | 529 | 63 | |
GenBank mRNA | AF145653.2 | - | 1 - 2 | 491 | 63 | |
BT021328.1 | - | 1 - 2 | 535 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |