Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104090.2 | NP_001097560.1 | 4 - 5 | 1425 | 65 | |
NM_001104091.2 | NP_001097561.1 | 3 - 4 | 1277 | 65 | ||
NM_001104092.2 | NP_001097562.1 | 2 - 3 | 224 | 65 | ||
NM_001300073.1 | NP_001287002.1 | 4 - 5 | 1425 | 65 | ||
NM_001300074.1 | NP_001287003.1 | 4 - 5 | 928 | 65 | ||
NM_140054.3 | NP_648311.2 | 3 - 4 | 780 | 65 | ||
NM_168343.2 | NP_729491.1 | 2 - 3 | 677 | 65 | ||
GenBank mRNA | BT029122.1 | - | 3 - 4 | 1245 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |