Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scramb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140132.3 | NP_648389.3 | 4 - 5 | 1399 | 73 | |
NM_168403.2 | NP_729600.1 | 3 - 4 | 990 | 73 | ||
NM_168404.2 | NP_729601.1 | 3 - 4 | 1003 | 73 | ||
GenBank mRNA | AY058486.1 | - | 4 - 5 | 1359 | 73 | |
BT089011.1 | - | 3 - 4 | 997 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |