Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JIL-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274768.1 | NP_001261697.1 | 5 - 6 | 2087 | 61 | |
NM_140175.3 | NP_648432.1 | 5 - 6 | 2087 | 61 | ||
NM_168439.2 | NP_729661.1 | 4 - 5 | 1249 | 61 | ||
GenBank mRNA | AF142061.1 | - | 5 - 6 | 2042 | 61 | |
BT044484.1 | - | 4 - 5 | 1241 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |