Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140289.3 | NP_648546.1 | 1 - 2 | 366 | 80 | |
GenBank mRNA | AY069574.1 | - | 1 - 2 | 310 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |