Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274840.1 | NP_001261769.1 | 4 - 5 | 1780 | 77 | |
NM_001274841.1 | NP_001261770.1 | 4 - 5 | 1792 | 77 | ||
NM_001274844.1 | NP_001261773.1 | 4 - 5 | 1780 | 77 | ||
NM_001274846.1 | NP_001261775.1 | 4 - 5 | 1792 | 77 | ||
NM_140328.5 | NP_648585.3 | 4 - 5 | 1780 | 77 | ||
GenBank mRNA | AY069066.1 | - | 4 - 5 | 1762 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |