Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sowah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168503.5 | NP_729816.4 | 8 - 9 | 1311 | 115 | |
GenBank mRNA | AY051801.1 | - | 8 - 9 | 1291 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |