Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169962.2 | NP_001163433.1 | 5 - 6 | 1228 | 68 | |
NM_140344.3 | NP_648601.1 | 5 - 6 | 1228 | 68 | ||
GenBank mRNA | AY129445.1 | - | 5 - 6 | 1197 | 68 | |
BT021340.1 | - | 5 - 6 | 1163 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |