Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144472.3 | NP_001137944.1 | 2 - 3 | 464 | 70 | |
NM_001144473.3 | NP_001137945.1 | 2 - 3 | 464 | 70 | ||
NM_001274856.2 | NP_001261785.1 | 2 - 3 | 464 | 70 | ||
NM_001274857.2 | NP_001261786.1 | 2 - 3 | 464 | 70 | ||
NM_001274858.2 | NP_001261787.1 | 2 - 3 | 464 | 70 | ||
NM_140350.5 | NP_648607.3 | 2 - 3 | 464 | 70 | ||
GenBank mRNA | BT126101.1 | - | 2 - 3 | 426 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |