Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: milt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164738.3 | NP_723251.1 | 1 - 2 | 618 | 61 | |
GenBank mRNA | AY051825.1 | - | 1 - 2 | 760 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |