Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nAChRalpha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201835.3 | NP_001188764.1 | 3 - 4 | 2235 | 73 | |
NM_001273373.2 | NP_001260302.1 | 3 - 4 | 2235 | 73 | ||
NM_135472.6 | NP_609316.4 | 2 - 3 | 1517 | 73 | ||
NM_164874.3 | NP_723494.2 | 3 - 4 | 2441 | 73 | ||
NM_205951.2 | NP_995673.1 | 2 - 3 | 1517 | 73 | ||
NM_205952.3 | NP_995674.1 | 3 - 4 | 2441 | 73 | ||
NM_205953.3 | NP_995675.1 | 2 - 3 | 1311 | 73 | ||
GenBank mRNA | AF321445.1 | - | 1 - 2 | 448 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |