Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164990.3 | NP_723721.1 | 1 - 2 | 370 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |