Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273130.1 | NP_001260059.1 | 7 - 8 | 2425 | 67 | |
NM_001273131.1 | NP_001260060.1 | 7 - 8 | 2353 | 67 | ||
NM_001298678.1 | NP_001285607.1 | 6 - 7 | 2152 | 67 | ||
NM_135031.4 | NP_608875.1 | 6 - 7 | 2137 | 67 | ||
NM_135032.2 | NP_608876.1 | 6 - 7 | 2070 | 67 | ||
NM_164603.2 | NP_723024.2 | 7 - 8 | 2281 | 67 | ||
NM_175966.2 | NP_787980.1 | 7 - 8 | 2365 | 67 | ||
NM_175967.2 | NP_787981.1 | 7 - 8 | 2403 | 67 | ||
NM_175968.2 | NP_787982.1 | 7 - 8 | 2349 | 67 | ||
GenBank mRNA | AF145605.1 | - | 6 - 7 | 2116 | 67 | |
AY050571.1 | - | 7 - 8 | 2368 | 67 | ||
AY050572.1 | - | 6 - 7 | 2119 | 67 | ||
BT016131.1 | - | 7 - 8 | 2368 | 67 | ||
BT044574.1 | - | 6 - 7 | 2065 | 67 | ||
BT126019.1 | - | 4 - 5 | 1588 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |