Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135067.3 | NP_608911.1 | 2 - 3 | 369 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |